missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Color Standard, APHA / Hazen (Platinum-Cobalt), Color = 5, Ricca Chemical

$165.21 - $228.91

Specifications

Chemical Name or Material Color Standard
Name Note Color = 5
CAS Min % 98.0,0.1,0.0012,9.0E-4
CAS Max % 100.0,0.11,0.0012,9.0E-4
CAS 7647-01-0,16921-30-5,7791-13-1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
2230005-16
View Documents
Ricca Chemical Company
RI2230.00516
500mL Glass amber
Each for $165.21
 
R2230005100
View Documents
Ricca Chemical Company
RIR2230005100C
100mL Glass amber N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
R22300054C
View Documents
Ricca Chemical Company
RIR22300054C
4L Glass amber
Each for $228.91
 
Description

Description

Specifications

Specifications

Color Standard
98.0,0.1,0.0012,9.0E-4
7647-01-0,16921-30-5,7791-13-1
Color = 5
100.0,0.11,0.0012,9.0E-4
Liquid
SDS