Please login to your online account to display your discounted pricing

Color Standard, APHA / Hazen (Platinum-Cobalt), Color = 400, Ricca Chemical

Chemical Identifiers

Chemical Name or Material Color Standard
Name Note Color = 400
CAS 7732-18-5
CAS Min % 93.0
CAS Max % 97.0
CAS 7647-01-0
CAS Min % 4.64
CAS Max % 4.83
CAS 16921-30-5
CAS Min % 0.05
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

2230400-16

 
ricca chemical company
RI2230.40016
Glass amber 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

R22304001C

 
ricca chemical company
RIR22304001C
Glass amber 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Color Clear yellow to yellow-brown
Physical Form Liquid
Quantity 500mL
Chemical Name or Material Color Standard
CAS Max % 0.05
CAS 7791-13-1
CAS Min % 0.04
CAS Max % 0.04
Name Note Color = 400
CAS 7732-18-5
CAS Min % 93.0
CAS Max % 97.0
CAS 7647-01-0
Packaging Glass amber
CAS Min % 4.64
CAS Max % 4.83
Methods APHA / Hazen (Platinum-Cobalt)
CAS 16921-30-5
CAS Min % 0.05