Please login to your online account to display your discounted pricing

Color Standard, APHA / Hazen (Platinum-Cobalt), Color = 300, Ricca Chemical

Chemical Identifiers

Chemical Name or Material Color Standard
Name Note Color = 300
CAS 7732-18-5
CAS Min % 94.0
CAS Max % 98.0
CAS 7647-01-0
CAS Min % 3.92
CAS Max % 4.08
CAS 16921-30-5
CAS Min % 0.04
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

2230300-16

  SDS
ricca chemical company
RI2230.30016
Glass amber 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

R22303001C

 
ricca chemical company
RIR22303001C
Glass amber 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Color Clear yellow to yellow-brown
Physical Form Liquid
Quantity 500mL
Methods APHA / Hazen (Platinum-Cobalt)
Chemical Name or Material Color Standard
CAS 7732-18-5
CAS Max % 0.04
CAS 7791-13-1
CAS Min % 0.03
CAS Max % 0.03
Name Note Color = 300
CAS Min % 94.0
CAS Max % 98.0
CAS 7647-01-0
Packaging Glass amber
CAS Min % 3.92
CAS Max % 4.08
CAS 16921-30-5
CAS Min % 0.04