missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Color Standard, APHA / Hazen (Platinum-Cobalt), Color = 30, Ricca Chemical

$422.76 - $422.76

Chemical Identifiers

CAS 7732-18-5,7647-01-0,16921-30-5,7791-13-1
CAS Min % 97.0,0.6,0.01,0.01
CAS Max % 100.0,0.62,0.01,0.01
Physical Form Liquid
Chemical Name or Material Color Standard, Color = 30
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
2230030-16 SDS Ricca Chemical Company
RI2230.03016
Glass amber 500mL Each for $422.76
R2230030100 Ricca Chemical Company
RIR2230030100C
Glass amber 100mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,7647-01-0,16921-30-5,7791-13-1
100.0,0.62,0.01,0.01
Color Standard, Color = 30
97.0,0.6,0.01,0.01
Liquid

Specifications

Liquid
500mL
APHA / Hazen (Platinum-Cobalt)
97.0,0.01,0.01,0.6
Color Standard, Color = 30
Clear light yellow to yellow brown
7732-18-5,7647-01-0,16921-30-5,7791-13-1
0.01,100.0,0.01,0.62
Glass amber