Color Standard, APHA/Hazen (Platinum-Cobalt), Color = 30, Ricca Chemical
Supplier: Ricca Chemical Company 223003016
Specifications
Color Standard | |
Amber Glass Bottle | |
97.0,0.6,0.01,0.01 | |
7647-01-0,16921-30-5,7791-13-1,7647-01-0,7732-18-5 | |
APHA / Hazen (Platinum-Cobalt) |
Color = 30 | |
500 mL | |
100.0,0.62,0.01,0.01 | |
Brown/Yellow to Yellow | |
Liquid |