missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Color Standard, APHA / Hazen (Platinum-Cobalt), Color = 200, Ricca Chemical

$582.52 - $582.52

Chemical Identifiers

CAS 7732-18-5,7647-01-0,16921-30-5,7791-13-1
CAS Min % 95.0,2.99,0.03,0.03
CAS Max % 99.0,3.11,0.03,0.03
Physical Form Liquid
Chemical Name or Material Color Standard, Color = 200
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
2230200-16 SDS Ricca Chemical Company
RI2230.20016
Glass amber 500mL Each for $582.52
R22302001C Ricca Chemical Company
RIR22302001C
Glass amber 1L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,7647-01-0,16921-30-5,7791-13-1
99.0,3.11,0.03,0.03
Color Standard, Color = 200
95.0,2.99,0.03,0.03
Liquid

Specifications

7732-18-5,7647-01-0,16921-30-5,7791-13-1
99.0,3.11,0.03,0.03
Glass amber
Clear yellow to yellow-brown
Color Standard, Color = 200
95.0,2.99,0.03,0.03
Liquid
APHA / Hazen (Platinum-Cobalt)
500mL