Color Standard, APHA / Hazen (Platinum-Cobalt), Color = 200, Ricca Chemical
Manufacturer:  Ricca Chemical Company RIR22302001C
 View more versions of this product
Catalog No. R22302001C
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications
Specifications
Color Standard
95.0,2.99,0.03,0.03
7647-01-0,16921-30-5,7791-13-1,7647-01-0,7732-18-5
Glass amber
Brown-Yellow to Yellow
Color = 200
99.0,3.11,0.03,0.03
Liquid
APHA / Hazen (Platinum-Cobalt)
1L
Documents