missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Color Standard, APHA / Hazen (Platinum-Cobalt), Color = 2, Ricca Chemical

$401.18 - $401.18

Chemical Identifiers

CAS 7732-18-5,7647-01-0,16921-30-5,7791-13-1
CAS Min % 98.0,0.03,4.0E-4,3.0E-4
CAS Max % 100.0,0.04,4.0E-4,3.0E-4
Physical Form Liquid
Chemical Name or Material Color Standard, Color = 2
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
2230002-16 SDS Ricca Chemical Company
RI2230.00216
Glass amber 500mL Each for $401.18
R2230002100 Ricca Chemical Company
RIR2230002100C
Glass amber 100mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,7647-01-0,16921-30-5,7791-13-1
100.0,0.04,4.0E-4,3.0E-4
Color Standard, Color = 2
98.0,0.03,4.0E-4,3.0E-4
Liquid

Specifications

7732-18-5,7647-01-0,16921-30-5,7791-13-1
100.0,0.04,4.0E-4,3.0E-4
Glass amber
Clear light yellow to yellow brown
Color Standard, Color = 2
98.0,0.03,4.0E-4,3.0E-4
Liquid
APHA / Hazen (Platinum-Cobalt)
500mL