missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Color Standard, APHA / Hazen (Platinum-Cobalt), Color = 15, Ricca Chemical

$422.45 - $422.45

Chemical Identifiers

CAS 7732-18-5,7647-01-0,16921-30-5,7791-13-1
CAS Min % 98.0,0.31,0.0033,0.0026
CAS Max % 100.0,0.32,0.0035,0.0028
Physical Form Liquid
Chemical Name or Material Color Standard, Color = 15
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
2230015-16 SDS Ricca Chemical Company
RI2230.01516
Amber Glass Bottle 500mL Each for $422.45
R2230015100 Ricca Chemical Company
RIR2230015100C
Amber Glass Bottle 100mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
R22300151C Ricca Chemical Company
RIR22300151C
Amber Glass Bottle 1L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,7647-01-0,16921-30-5,7791-13-1
100.0,0.32,0.0035,0.0028
Color Standard, Color = 15
98.0,0.31,0.0033,0.0026
Liquid

Specifications

Liquid
500mL
Amber Glass Bottle
0.31,98.0,0.0026,0.0033
Color Standard, Color = 15
Clear light yellow to yellow brown
APHA / Hazen (Platinum-Cobalt)
16921-30-5,7732-18-5,7791-13-1,7647-01-0
0.32,0.0028,0.0035,100.0