Please login to your online account to display your discounted pricing

Color Standard, APHA / Hazen (Platinum-Cobalt), Color = 100, Ricca Chemical


Quantity 1L
Packaging Glass amber

Manufacturer: ricca chemical company  RIR22301001C

 View more versions of this product

Catalog No. R22301001C

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Color Standard
Name Note Color = 100
CAS 7791-13-1 (Cobalt(II) Chloride, Hexahydrate)
CAS 7647-01-0 (Hydrochloric Acid)
CAS 16921-30-5 (Potassium Hexachloroplatinate (IV))
CAS 7732-18-5 (Water, Deionized)
Physical Form Liquid
Packaging Glass amber
Methods APHA / Hazen (Platinum-Cobalt)
Color Clear light yellow to yellow brown
Quantity 1L

Specifications

Chemical Name or Material Color Standard
Name Note Color = 100
CAS 7791-13-1 (Cobalt(II) Chloride, Hexahydrate)
CAS 7647-01-0 (Hydrochloric Acid)
CAS 16921-30-5 (Potassium Hexachloroplatinate (IV))
CAS 7732-18-5 (Water, Deionized)
Physical Form Liquid