Color Standard, APHA/Hazen (Platinum-Cobalt), Color=10, Ricca Chemical
Platinum-Cobalt (APHA/Hazen) Color Standards
Manufacturer:  Ricca Chemical Company RIR22305001C
 View more versions of this product
Catalog No. R22305001C
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
Specifications
Specifications
Color Standard, APHA/Hazen (Platinum-Cobalt), Color=10
99.91,0.09,0.0026,0.002
Miscible in water
Glass Amber Bottle
<1
100°C
1L
99.91,0.09,0.0026,0.002
16921-30-5,7791-13-1,7647-01-0,7732-18-5
Liquid
1.001
0°C
Brown-Yellow