missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Color Standard, APHA/Hazen (Platinum-Cobalt), Color=10, Ricca Chemical

Platinum-Cobalt (APHA/Hazen) Color Standards

Specifications

CAS 16921-30-5,7791-13-1,7647-01-0
CAS Max % 99.91,0.09,0.0026,0.002
CAS Min % 99.91,0.09,0.0026,0.002
Chemical Name or Material Color Standard, APHA/Hazen (Platinum-Cobalt), Color=10
Density 1.001
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
R22305001C
View Documents
Ricca Chemical Company
RIR22305001C
1L N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
R2230500250
View Documents
Ricca Chemical Company
RIR2230500250C
250mL N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Specifications

16921-30-5,7791-13-1,7647-01-0
99.91,0.09,0.0026,0.002
1.001
Brown-Yellow
<1
Liquid
99.91,0.09,0.0026,0.002
Color Standard, APHA/Hazen (Platinum-Cobalt), Color=10
100°C
0°C
Glass Amber Bottle
Miscible in water
SDS