missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Color Standard, APHA / Hazen (Platinum-Cobalt), Color = 10, Ricca Chemical

$384.36 - $384.36

Chemical Identifiers

CAS 7732-18-5,7647-01-0,16921-30-5,7791-13-1
CAS Min % 98.0,0.21,0.0023,0.0018
CAS Max % 100.0,0.21,0.0023,0.0018
Physical Form Liquid
Chemical Name or Material Color Standard
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
22305-16 Ricca Chemical Company
RI2230.516
Glass amber 500mL Each for $384.36
R2230500100 Ricca Chemical Company
RIR2230500100C
Glass amber 100mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
R22305004C Ricca Chemical Company
RIR22305004C
Glass amber 4L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,7647-01-0,16921-30-5,7791-13-1
100.0,0.21,0.0023,0.0018
Color Standard
98.0,0.21,0.0023,0.0018
Liquid

Specifications

7732-18-5,7647-01-0,16921-30-5,7791-13-1
100.0,0.21,0.0023,0.0018
Glass amber
Clear light yellow to yellow brown
Color Standard
98.0,0.21,0.0023,0.0018
Liquid
APHA / Hazen (Platinum-Cobalt)
500mL

Safety and Handling

ShelfLife : 12 months