Please login to your online account to display your discounted pricing

Color Reagent, for Nitrate Determination, Ricca Chemical

Chemical Identifiers

Chemical Name or Material Color Reagent
CAS 7732-18-5
CAS Min % 81.0
CAS Max % 84.0
CAS 7664-38-2
CAS Min % 13.68
CAS Max % 14.23
CAS 63-74-1
CAS Min % 3.24
CAS Max % 3.37
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

22331

 
ricca chemical company
RI22331
Glass amber 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

2233-16

 
ricca chemical company
RI223316
Glass amber 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

2233-4

 
ricca chemical company
RI22334
Glass amber 120mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

223332

 
ricca chemical company
RI223332
Glass amber 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless
Quantity 4L
Chemical Name or Material Color Reagent
CAS 7732-18-5
CAS Max % 3.37
CAS 1465-25-4
CAS Min % 0.16
CAS Max % 0.17
CAS Min % 81.0
CAS Max % 84.0
CAS 7664-38-2
Packaging Glass amber
CAS Min % 13.68
CAS Max % 14.23
CAS 63-74-1
CAS Min % 3.24

For Nitrate Determination