Please login to your online account to display your discounted pricing

Color Reagent for Chloride, Certified, LabChem™

$72.97 - $253.23

Chemical Identifiers

Chemical Name or Material Color Reagent for Chloride
CAS 7732-18-5
CAS Max % 81.605
CAS 67-56-1
CAS Max % 14.937
CAS 7782-61-8
CAS Max % 3.03
CAS 7697-37-2
CAS Max % 0.315
CAS 592-85-8
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC132601

 
lab chem
LC132601
500mL Each for $72.97

LC132602

 
lab chem
LC132602
1L Each for $103.97

LC132604

 
lab chem
LC132604
4L Each for $253.23
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 404.02
Chemical Name or Material Color Reagent for Chloride
CAS Max % 3.03
Decomposition Information Mercury; Carbon monoxide; Carbon dioxide; Sulfur compounds; Nitrogen oxides
CAS 7697-37-2
Color Clear orange
Quantity 500mL
CAS Max % 0.315
CAS 592-85-8
CAS Max % 0.063
Physical Form Liquid
Linear Formula Fe(NO3)3·9H2O
CAS 7732-18-5
Health Hazard 1 Danger
CAS Max % 81.605
Health Hazard 2 GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Suspected of damaging fertility or the unborn child.
Causes damage to organs (central nervous system, optic nerve).
Harmful to aquatic life with long lasting effects.
Identification Passes Test
CAS 67-56-1
Health Hazard 3 GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Do not breathe mist, vapors, spray.
Do not eat, drink or smoke when using this product.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If exposed or concerned: Get medical advice/attention.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Packaging Amber Glass
CAS Max % 14.937
Solubility Soluble in water
CAS 7782-61-8
Suitability Passes Test
Storage Information Room Temperature
Safety and Handling

Danger

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Do not breathe mist, vapors, spray.
Do not eat, drink or smoke when using this product.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If exposed or concerned: Get medical advice/attention.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Suspected of damaging fertility or the unborn child.
Causes damage to organs (central nervous system, optic nerve).
Harmful to aquatic life with long lasting effects.