Color Filter Sets
Explore the science of colors.
Manufacturer:  Fisher Science Education IFFS4
 View more versions of this product
Catalog No. S63192
Description
Description
Variety of colors individually mounted in 2 × 2 in. (5 × 5cm) 35mm plastic slide holders.
Basic Color Filter Set:
  • Three filters: red, blue, green
  • Advanced Color Filter Set:
    • 14 filters: light blue, medium blue, daylight blue, dark blue, golden amber, medium green, light green, green-blue, light red, medium purple, orange, medium yellow, medium red, frost
    Specifications
    Specifications
    Advanced color filter set
    Ideal for use in optical transmission measurements, solar monitoring and exploration, attenuation of laser beams and demonstrations of laser light's monochromacity
    14 Filters: light blue, medium blue, daylight blue, dark blue, golden amber, medium green, light green, green-blue, light red, medium purple, orange, medium yellow, medium red, frost