Color-Coded Spring Scales
Clear plastic tube allows students to see the action of the spring.
Manufacturer:  Fisher Science Education 39650
 View more versions of this product
Catalog No. S02324
Specifications
Description
Scales, Spring; SI Manufacturing; Capacity: 2kg/20 Newtons; Color-coded: Red
Series of eight color-coded tubular spring scales can be used for both force and mass measurements.
  • Calibrated in both Newtons and grams with a beveled top for use on an inclined plane
  • Incorporates a unique zero adjustment
Description & Specifications
Specifications
4.4 lb., 2kg
2kg
4.4 lb.
Red
4+
Spring Scale