Please login to your online account to display your discounted pricing

Color-Coded Spring Scales


Clear plastic tube allows students to see the action of the spring.

Manufacturer: fisher science education  39600/6

 View more versions of this product

Catalog No. S02321A

  • / Pack of 6 Each
 

Description & Specifications

Specifications

Includes Set of 6 colors.
Type Spring Scale
Color Multi
Grade 4+

Series of eight color-coded tubular spring scales can be used for both force and mass measurements.

  • Calibrated in both Newtons and grams with a beveled top for use on an inclined plane
  • Incorporates a unique zero adjustment