Please login to your online account to display your discounted pricing
American Educational Products Color-Coded Spring Scales
Clear plastic tube allows students to see the action of the spring.
$3.05 - $46.80
Specifications
Type Spring Scale
View More Specs
Products
Catalog Number Mfr. No. Capacity Color Price Quantity    
S40142A
american educational products
03960
250g/2.5N Blue Each for $5.70
Case of 100 Each for $567.00
S40142B
american educational products
03961
500g/5N Green Each for $5.70
Case of 100 Each for $567.00
S40142C
american educational products
03962
1000g/10N Beige Each for $5.70
Case of 100 Each for $567.00
S40142F
american educational products
03965
5000g/50N Yellow Each for $3.05
Case of 100 Each for $303.75
S40142E
american educational products
03964
3000g/30N White Each for $3.05
Case of 100 Each for $303.75
S40142G
fisher science education
S40142G
Set of 6: 250g, 500g, 1000g, 2000g, 3000g, 5000g Multi colors Each for $46.80
Description & Specifications
Specifications
Type Spring Scale
Series of six color-coded tubular spring scales can be used for both force and mass measurements.
  • Calibrated in both Newtons and grams with a beveled top for use on an inclined plane
  • Incorporate a unique zero adjustment with a clear plastic tube, allowing students to see spring action
Available separately or as a set of all six colors