Please login to your online account to display your discounted pricing

Collodion; USP; Flexible; J.T.Baker™

Chemical Identifiers

Chemical Name or Material Collodion
CAS 8001-79-4
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MK4580150

 
avantor performance
458002
Glass Bottle 150mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 150mL
Chemical Name or Material Collodion
CAS 8001-79-4
Packaging Glass Bottle