Please login to your online account to display your discounted pricing

Collodion; USP; Flexible; J.T.Baker™


Quantity 150mL
Packaging Glass Bottle

Manufacturer: avantor performance  458002

 View more versions of this product

Catalog No. MK4580150

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Quantity 150mL
Chemical Name or Material Collodion
Packaging Glass Bottle
CAS 8001-79-4

Specifications

Chemical Name or Material Collodion
CAS 8001-79-4