missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Collagen X Ab-1, Mouse Monoclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry and immunofluorescence experiments with Thermo Scientific™ Collagen X Ab-1, Mouse Monoclonal Antibody.

$463.31 - $2,016.24

Specifications

Antigen Collagen X Ab-1
Clone X-AC9
Applications Immunofluorescence,Immunohistochemistry (Paraffin),Immunofluorescence,Immunohistochemistry (Paraffin)
Classification Monoclonal
Format Concentrated
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Concentration Conjugate Quantity Price Quantity  
Catalog Number Mfr. No. Concentration Conjugate Quantity Price Quantity  
MS852B Thermo Scientific™ Lab Vision™
MS852B
200μg/mL Biotin 1mL Each for $2,016.24
MS852B0 Thermo Scientific™ Lab Vision™
MS852B0
200μg/mL Biotin 100μL Each for $644.63
MS852B1 Thermo Scientific™ Lab Vision™
MS852B1
200μg/mL Biotin 500μL Each for $1,494.80
MS852P Thermo Scientific™ Lab Vision™
MS852P
200μg/mL Unlabeled 1mL Each for $1,597.27
MS852P0 Thermo Scientific™ Lab Vision™
MS852P0
200μg/mL Unlabeled 100μL Each for $463.31
MS852P1 Thermo Scientific™ Lab Vision™
MS852P1
200μg/mL Unlabeled 500μL Each for $1,161.76
MS852P1ABX Thermo Scientific™ Lab Vision™
MS852P1ABX
1mg/mL Unlabeled 100μL Each for $1,289.39
MS852PABX Thermo Scientific™ Lab Vision™
MS852PABX
1mg/mL Unlabeled 200μL Each for $1,755.73
Specifications

Description

Obtain accurate, reproducible results in immunohistochemistry and immunofluorescence experiments with Thermo Scientific™ Collagen X Ab-1, Mouse Monoclonal Antibody.

Type X collagen is a non-fibrillar, short-chain, network-forming collagen found in fetal hypertrophic cartilage in the growth zones of long bones, vertebrae, and ribs. It is a short collagen with a triple helical core half the length of the fibril-forming collagen type I, II, and III and a globular domain at the carboxyl end (NC-1 domain) through which the molecules assemble into an hexagonal meshwork. The function of collagen X, a unique homotrimer synthesised by hypertrophic chondrocytes, is not known but its localization and transient expression at sites of calcification suggest that it is likely to be associated with events in the early stages of endochondral bone formation.

Host Species: Mouse

Clone: X-AC9

Isotype: IgG1/κ

Species Reactivity: Chicken. Does not react with human. Others not known.

Epitope: Not determined

Immunogen: Purified chicken type X collagen

Positive Control: Chicken cartilage

Cellular Localization: Cartilage matrix

Recommended for:

  • Immunohistochemistry (Frozen and formalin/paraffin; not qualified for IHC on human tissues)
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

Collagen X Ab-1
Immunofluorescence,Immunohistochemistry (Paraffin),Immunofluorescence,Immunohistochemistry (Paraffin)
Concentrated
RUO
Primary
IgG2a kappa
X-AC9
Monoclonal
Murine
Extracellular Matrix
Purified chicken type X collagen