Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Collagen X Ab-1, Mouse Monoclonal Antibody


Obtain accurate, reproducible results in immunohistochemistry and immunofluorescence experiments with Thermo Scientific™ Collagen X Ab-1, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS852P1ABX

 View more versions of this product

Catalog No. MS852P1ABX

  • / Each
 

Description & Specifications

Specifications

Antigen Collagen X Ab-1
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone X-AC9
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Chicken
Format Concentrated
Host Species Murine
Immunogen Purified chicken type X collagen
Isotype IgG2a kappa
Quantity 100μL
Regulatory Status RUO
Research Discipline Extracellular Matrix
Type Primary
Monoclonal or Polyclonal Monoclonal
Target Species Chicken

Obtain accurate, reproducible results in immunohistochemistry and immunofluorescence experiments with Thermo Scientific™ Collagen X Ab-1, Mouse Monoclonal Antibody.

Type X collagen is a non-fibrillar, short-chain, network-forming collagen found in fetal hypertrophic cartilage in the growth zones of long bones, vertebrae, and ribs. It is a short collagen with a triple helical core half the length of the fibril-forming collagen type I, II, and III and a globular domain at the carboxyl end (NC-1 domain) through which the molecules assemble into an hexagonal meshwork. The function of collagen X, a unique homotrimer synthesised by hypertrophic chondrocytes, is not known but its localization and transient expression at sites of calcification suggest that it is likely to be associated with events in the early stages of endochondral bone formation.

Host Species: Mouse

Clone: X-AC9

Isotype: IgG1/κ

Species Reactivity: Chicken. Does not react with human. Others not known.

Epitope: Not determined

Immunogen: Purified chicken type X collagen

Positive Control: Chicken cartilage

Cellular Localization: Cartilage matrix

Recommended for:

  • Immunohistochemistry (Frozen and formalin/paraffin; not qualified for IHC on human tissues)
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO