missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Collagen VII Ab-1, Mouse Monoclonal Antibody

Ensure accurate and reproducible results in immunohistochemistry, immunofluorescence, and electron microscopy experiments.

$238.02 - $929.88

Specifications

Antigen Collagen VII Ab-1
Clone LH7.2
Applications Immunofluorescence,Immunohistochemistry (Frozen),Immunofluorescence,Immunohistochemistry (Frozen)
Classification Monoclonal
Conjugate Unlabeled
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Concentration Quantity Price Quantity  
Catalog Number Mfr. No. Concentration Quantity Price Quantity  
MS444P Thermo Scientific™ Lab Vision™
MS444P
200μg/mL 1mL Each for $822.82
MS444P0 Thermo Scientific™ Lab Vision™
MS444P0
200μg/mL 100μL Each for $238.02
MS444P1 Thermo Scientific™ Lab Vision™
MS444P1
200μg/mL 500μL Each for $651.43
MS444P1ABX Thermo Scientific™ Lab Vision™
MS444P1ABX
1mg/mL 100μL Each for $747.10
MS444PABX Thermo Scientific™ Lab Vision™
MS444PABX
1mg/mL 200μL Each for $929.88
Specifications

Description

Ensure accurate and reproducible results in immunohistochemistry, immunofluorescence, and electron microscopy experiments with Thermo Scientific™ Collagen VII Ab-1, Mouse Monoclonal Antibody.

Host Species: Mouse

Clone: LH7.2

Isotype: IgG1/κ

Species Reactivity: Human, Marmoset, Monkey, Cow, Sheep, Goat, Pig and Guinea Pig.

Epitope: On nonhelical (noncollagenous) C-terminal region of collagen VII and is resistant to collagenase, trypsin, pronase, neuraminidase, chondroitinase and hyaluronidase. However, it is lost by digestion of collagen VII with pepsin.

Immunogen: Insoluble fractions of human neonatal foreskin epidermal cells

Molecular Weight: 250 kDa (intact) 150 kDa (digested)

Positive Control: Normal skin

Cellular Localization: Epidermal basement membrane

Recommended for:

  • Immunohistochemistry (Acetone-fixed frozen only; fixation in methanol is not suitable)
  • Immunofluorescence
  • Electron Microscopy

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

Collagen VII Ab-1
Immunofluorescence,Immunohistochemistry (Frozen),Immunofluorescence,Immunohistochemistry (Frozen)
Unlabeled
Murine
Extracellular Matrix
Insoluble fractions of human neonatal foreskin epidermal cells
LH7.2
Monoclonal
Concentrated
RUO
Primary
IgG2a kappa