Please login to your online account to display your discounted pricing

Collagen Type X Mouse, eFluor 570, Clone: X53, eBioscience™

Mouse Monoclonal Antibody

$205.90 - $487.20

Specifications

Antigen Collagen Type X
Clone X53
Host Species Mouse
Species Reactivity Canine, Guinea Pig, Human, Mouse, Pig, Rat
Applications Immunocytochemistry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123651

 
life technologies
LS41977180
25μg Each for $205.90

501123652

 
life technologies
LS41977182
100μg Each for $487.20
Description & Specifications

Specifications

Antigen Collagen Type X
Clone X53
Host Species Mouse
Species Reactivity Canine, Guinea Pig, Human, Mouse, Pig, Rat
Applications Immunocytochemistry
Applications Immunohistochemistry (Formalin/Paraffin)
Applications Immunohistochemistry (Frozen)
Applications Microscopy
Regulatory Status RUO
Conjugate eFluor 570
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The monoclonal antibody X53 recognizes human, mouse, porcine, guinea pig, and rat Collagen Type X. X53 does not cross-react with collagen types I, II, II, IV, or VI. Collagen Type X is a homotrimeric short chain collagen. Collagen Type X has limited expression in normal adult tissues, primarily being expressed during bone growth and development. Expression is localized to the peri- and extracellular matrix of hypertrophic chondrocytes during endochondral ossification and also plays a role in cartilage mineralization. Collagen Type X is elevated in many tumor types with expression being localized in the vasculature. Mutations to the gene COL10A1, which encodes human Collagen Type X, is associated with Schmid type metaphyseal chondrodysplasia.