Please login to your online account to display your discounted pricing

Collagen Type IV Mouse anti-Human, eFluor 660, Clone: 1042, eBioscience™

Mouse Monoclonal Antibody

$155.15 - $375.55

Specifications

Antigen Collagen Type IV
Clone 1042
Host Species Mouse
Species Reactivity Human
Applications Immunohistochemistry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501124574

 
life technologies
LS50987180
25μg Each for $155.15

501124575

 
life technologies
LS50987182
100μg Each for $375.55
Description & Specifications

Specifications

Antigen Collagen Type IV
Clone 1042
Host Species Mouse
Species Reactivity Human
Applications Immunohistochemistry
Applications Immunohistochemistry (Formalin/Paraffin)
Applications Immunohistochemistry (Frozen)
Regulatory Status RUO
Conjugate eFluor 660
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

This monoclonal antibody 1042 recognizes human collagen Type IV. Collagens are the main components of the extracellular matrix and provide the tensile strength to tissues. There are four main types of collagen (I, II, III and IV) which contribute to the connective tissue. Expression of type IV is restricted to the basal lamina. It contains two alpha-1 and one alpha-2-like chains. Collagen Type IV is associated with disorders such as Alport syndrome and Goodpasture's syndrome.