missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Collagen IV Mouse anti-Human, eFluor® 660, Clone: 1042, eBioscience™

Mouse Monoclonal Antibody

Manufacturer:  Life Technologies LS50987182

Catalog No. 501124575



Description

Description

Description: This monoclonal antibody 1042 recognizes human collagen Type IV. Collagens are the main components of the extracellular matrix and provide the tensile strength to tissues. There are four main types of collagen (I, II, III and IV) which contri

Collagen IV is a major constituent of the basement membranes along with laminins and enactins. It is composed of alpha 1 IV chain and alpha 2 IV chain in 2:1 ratio . It can form insoluble fibers with high tensile strength. Antibody to collagen IV is useful in detecting the loss of parts of basement membrane in carcinomas.
Specifications

Specifications

Collagen IV
Monoclonal
0.2 mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P02462
COL4A1
IgG2b, kappa
100 μg
4° C, store in dark, DO NOT FREEZE!
Primary
1282
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
1042
eFluor® 660
COL4A1
Liquid
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Documents

For Research Use Only.