missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Collagen IV, Alexa Fluor 488, clone: 1042, eBioscience™

Mouse Monoclonal Antibody

Manufacturer:  Life Technologies LS53987180

 View more versions of this product

Catalog No. 501124660



Description

Description

Collagen IV is a major constituent of the basement membranes along with laminins and enactins. It is composed of alpha 1 IV chain and alpha 2 IV chain in 2:1 ratio . It can form insoluble fibers with high tensile strength. Antibody to collagen IV is useful in detecting the loss of parts of basement membrane in carcinomas.

Description: This monoclonal antibody 1042 recognizes human collagen Type IV. Collagens are the main components of the extracellular matrix and provide the tensile strength to tissues. There are four main types of collagen (I, II, III and IV) which contribute to the connective tissue. Expression of type IV is restricted to the basal lamina. It contains two alpha-1 and one alpha-2-like chains. Collagen Type IV is associated with disorders such as Alport syndrome and Goodpasture's syndrome.Applications Reported: This 1042 antibody has been reported for use in immunohistochemical staining of frozen tissue sections, and immunohistochemical staining of formalin-fixed paraffin embedded tissue sections.Applications Tested: This 1042 antibody has been tested by immunohistochemistry of formalin-fixed paraffin embedded human tissue using low pH antigen retrieval and can be used at less than or equal to 10 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

Collagen IV
Monoclonal
0.5mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P02462
COL4A1
IgG2b, kappa, kappa
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
1282
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
1042
Alexa Fluor 488
COL4A1
Liquid
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Documents

For Research Use Only.