Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Collagen IV Ab-2, Mouse Monoclonal Antibody

Ensure accurate and reproducible results in immunohistochemistry applications with Thermo Scientific™ Collagen IV Ab-2, Mouse Monoclonal Antibody.

$240.37 - $996.06

Specifications

Antigen Collagen IV Ab-2
Clone CIV 22
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

MS448S

 
thermo scientific™ lab vision™
MS448S
Culture Supernatant with Azide 1mL Each for $996.06

MS448S0

 
thermo scientific™ lab vision™
MS448S0
Culture Supernatant with Azide 100µL Each for $240.37

MS448S1

 
thermo scientific™ lab vision™
MS448S1
Culture Supernatant with Azide 500µL Each for $667.27
Description & Specifications

Specifications

Antigen Collagen IV Ab-2
Clone CIV 22
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Porcine
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Porcine
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Porcine
Format Concentrated
Host Species Mouse
Regulatory Status RUO
Research Discipline Extracellular Matrix
Type Primary
Formulation Culture Supernatant with Azide
Immunogen Human glomeruli
Isotype IgG1
Purification Method Skin

Ensure accurate and reproducible results in immunohistochemistry applications with Thermo Scientific™ Collagen IV Ab-2, Mouse Monoclonal Antibody.

Collagen IV is a major constituent of the basement membranes along with laminins and enactins. It is composed of alpha1(IV) chain and alpha2(IV) chain in 2:1 ratio . It can form insoluble fibers with high tensile strength. Antibody to collagen IV is useful in detecting the loss of parts of basement membrane in carcinomas.

Host Species: Mouse

Clone: CIV 22

Isotype: IgG1

Species Reactivity: Human and Cow. Does not react with rat, pig and dog. Others not known

Epitope: Not determined

Immunogen: Human glomeruli

Positive Control: Skin

Cellular Localization: Basement membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO