missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Collagen IV Ab-1, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ Collagen IV Ab-1, Mouse Monoclonal Antibody.

$243.18 - $761.55

Specifications

Antigen Collagen IV Ab-1
Clone PHM-12
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
MS375R7 Thermo Scientific™ Lab Vision™
MS375R7
7mL Each for $479.39
MS375S Thermo Scientific™ Lab Vision™
MS375S
1mL Each for $761.55
MS375S0 Thermo Scientific™ Lab Vision™
MS375S0
100μL Each for $243.18
MS375S1 Thermo Scientific™ Lab Vision™
MS375S1
500μL Each for $586.73
Specifications

Description

Ensure accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ Collagen IV Ab-1, Mouse Monoclonal Antibody.

Collagen IV is a major constituent of the basement membranes along with laminins and enactins. It is composed of α1(IV) chain and α2(IV) chain in 2:1 ratio . It can form insoluble fibers with high tensile strength. Antibody to collagen IV is useful in detecting the loss of parts of basement membrane in carcinomas.

Host Species: Mouse

Clone: PHM-12

Isotype: IgG1

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Human glomeruli

Positive Control: Skin

Cellular Localization: Basement membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

Collagen IV Ab-1
Immunohistochemistry (Paraffin)
Unlabeled
RUO
Primary
IgG1
PHM-12
Monoclonal
Murine
Extracellular Matrix
Human glomeruli