Learn More
Collagen II Ab-2 Mouse Monoclonal Antibody, Epredia™
Ensure accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence and flow cytometry experiments.
$498.33 - $1856.13
Specifications
Antigen | Collagen II Ab-2 |
---|---|
Clone | 2B1.5 |
Applications | Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Host Species | Mouse |
Catalog Number | Mfr. No. | Form | Conjugate | Quantity | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Form | Conjugate | Quantity | Price | Quantity | |||
MS235B0
|
Thermo Fisher Scientific
MS235B0 |
Concentrated | Biotin | 100 μL |
Each for $498.33
|
|
|||
MS235B1
|
Thermo Fisher Scientific
MS235B1 |
Concentrated | Biotin | 500 μL |
Each for $1,413.00
|
|
|||
MS235PABX
|
Thermo Fisher Scientific
MS235PABX |
Concentrated | Unconjugated | 200 μL |
Each for $1,856.13
|
|
|||
MS235P1
|
Thermo Fisher Scientific
MS235P1 |
Concentrated | Unconjugated | 500 μL |
Each for $1,094.63
|
|
|||
MS235P
|
Thermo Fisher Scientific
MS235P |
Concentrated | Unconjugated | 1 mL |
Each for $1,740.52
|
|
|||
Description
Host Species: Mouse
Clone: 2B1.5
Isotype: IgG2a/κ
Species Reactivity: Human, Mouse, Rat, Cow and Chicken. Others not known.
Epitope: Carboxyl-terminal one quarter of the type II collagen. The epitope for 2B1.5 MAb is not susceptible to digestion of lathyritic type II collagen with recombinant stromelysin
Immunogen: A purified preparation of lathyritic type II collagen from embryonic chicken sternum
Positive Control: Cartilage in lung or fetus
Cellular Localization: Cartilage matrix
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
- Flow Cytometry
Note:
USA: RUO; Int'l: RUO
Specifications
Collagen II Ab-2 | |
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Mouse | |
Extracellular Matrix | |
A purified preparation of lathyritic type II collagen from embryonic chicken sternum | |
Primary |
2B1.5 | |
Monoclonal | |
RUO | |
Bovine, Chicken, Human, Mouse, Rat | |
IgG2a κ |