missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Collagen II Ab-1, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in western blotting and immunofluorescence experiments with Thermo Scientific™ Collagen II Ab-1, Mouse Monoclonal Antibody.

$274.29 - $1,206.57

Specifications

Antigen Collagen II Ab-1
Clone 5B2.5
Applications Immunofluorescence,Western Blotting,Immunofluorescence,Western Blotting
Classification Monoclonal
Conjugate Unlabeled
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Concentration Quantity Price Quantity  
Catalog Number Mfr. No. Concentration Quantity Price Quantity  
MS234P Thermo Scientific™ Lab Vision™
MS234P
200μg/mL 1mL Each for $1,122.57
MS234P0 Thermo Scientific™ Lab Vision™
MS234P0
200μg/mL 100μL Each for $274.29
MS234P1 Thermo Scientific™ Lab Vision™
MS234P1
200μg/mL 500μL Each for $735.88
MS234P1ABX Thermo Scientific™ Lab Vision™
MS234P1ABX
1mg/mL 100μL Each for $825.10
MS234PABX Thermo Scientific™ Lab Vision™
MS234PABX
1mg/mL 200μL Each for $1,206.57
Specifications

Description

Ensure accurate, reproducible results in western blotting and immunofluorescence experiments with Thermo Scientific™ Collagen II Ab-1, Mouse Monoclonal Antibody.

Host Species: Mouse

Clone: 5B2.5

Isotype: IgG2a/κ

Species Reactivity: Human, Chicken and Mouse. Others not known.

Epitope: GGFDEK, slightly preceding the lysine residue involved in crosslinking. Two terminal glycine residues constitute a key structural element of the epitope. The epitope is lost after digestion of lathyritic type II collagen with recombinant stromelysin.

Immunogen: A purified preparation of lathyritic type II collagen from embryonic chicken sternum

Positive Control: Cartilage

Cellular Localization: Cartilage matrix

Recommended for:

  • Immunohistochemistry (Not suitable)
  • Western Blotting
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

Collagen II Ab-1
Immunofluorescence,Western Blotting,Immunofluorescence,Western Blotting
Unlabeled
Murine
Extracellular Matrix
A purified preparation of lathyritic type II collagen from embryonic chicken sternum
5B2.5
Monoclonal
Concentrated
RUO
Primary
IgG2a kappa