missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Colistin Sulfate (mixture), TCI America™

Manufacturer:  TCI America C29305G

 View more versions of this product

Catalog No. C29305G



Specifications

Specifications

Colistin Sulfate (mixture)
5g
MFCD00146495
Crystal-Powder at 20°C
White-Yellow
1264-72-8
2811
≥90.0% (HPLC)

Chemical Identifiers

1264-72-8
MFCD00146495
Safety and Handling

Safety and Handling

RTECSNumber : TR1500000

TSCA : No

Documents