missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Colistin Sulfate (mixture), TCI America™

$130.03 - $450.97

Chemical Identifiers

CAS 1264-72-8
MDL Number MFCD00146495
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C29305G View Documents Tci America
C29305G
5g Each for $130.03
 
C293025G View Documents Tci America
C293025G
25g Each for $450.97
 
Specifications

Chemical Identifiers

1264-72-8
MFCD00146495

Specifications

White-Yellow
1264-72-8
2811
≥90.0% (HPLC)
5g
MFCD00146495
Crystal-Powder at 20°C
Colistin Sulfate (mixture)
Safety and Handling

Safety and Handling

RTECSNumber : TR1500000

TSCA : No