Please login to your online account to display your discounted pricing

Alfa Aesar™ Colistin sulfate, 50mg/mL in distilled water, sterile-filtered

$52.74 - $52.74

Chemical Identifiers

Chemical Name or Material Colistin sulfate
CAS 1264-72-8
Name Note 50mg/mL in distilled water, sterile-filtered
MDL Number MFCD00146495
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ674158EQ

  SDS
alfa aesar™
J674158EQ
5 x 1mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67415XF

  SDS
alfa aesar™
J67415XF
1mL Each for $52.74
Description & Specifications

Specifications

Chemical Name or Material Colistin sulfate
MDL Number MFCD00146495
Health Hazard 2 GHS H Statement
H302
Harmful if swallowed.
Physical Form Liquid
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Storage Note 1 -30° to -10°C
Name Note 50mg/mL in distilled water, sterile-filtered
RTECS Number TR1500000
Quantity 5 x 1mL
CAS 1264-72-8
Safety and Handling

Warning

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302
Harmful if swallowed.

-30° to -10°C