Please login to your online account to display your discounted pricing

Alfa Aesar™ Colistin sodium methanesulfonate


Quantity 1g

Manufacturer: alfa aesar™  J6669803

 View more versions of this product

Catalog No. AAJ6669803

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 1759.89
CAS 8068-28-8
Chemical Name or Material Colistin sodium methanesulfonate
Synonym Colimycin sodium methanesulfonate; Polymyxin E sodium methanesulfonate
MDL Number MFCD00130824
Physical Form Powder
Solubility Information Freely soluble in water
RTECS Number GH1650000
Storage Note 1 2° to 8°C
Quantity 1g
TSCA TSCA
Merck Index 14,2479
Color White to pale yellow

Specifications

Chemical Name or Material Colistin sodium methanesulfonate
CAS 8068-28-8
Formula Weight 1759.89
MDL Number MFCD00130824
Merck Index 14,2479
Synonym Colimycin sodium methanesulfonate; Polymyxin E sodium methanesulfonate

Solubility
Freely soluble in water

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.

Accessories

2° to 8°C

SDS & Safety Documents