Cole-Parmer™ Masterflex™ Norprene™L/S™ High Performance Precision Pump Tubing
Use this tubing for pressure/vacuum applications. Cole Parmer™ Masterflex™ Norprene™ L/S™ High Performance Precision Pump Tubing is ideal for applications requiring excellent chemical, heat, ozone and ultraviolet (UV) light resistance.
Manufacturer:  Cole-Parmer™ 0640415
 View more versions of this product
Catalog No. MFX0640415
Description
Description
Use this tubing for pressure/vacuum applications. Cole Parmer™ Masterflex™ Norprene™ L/S™ High Performance Precision Pump Tubing is ideal for applications requiring excellent chemical, heat, ozone and ultraviolet (UV) light resistance.
Choose High Performance tubing for applications involving pressure, suction lift, viscous fluids or long tubing life.
  • Thicker wall compared to Precision pump tubing
  • Nonconductive and heat-sealable
  • Non-aging, non-oxidizing, with superior acid and alkali resistance
  • Opaque black
  • Complies with NSF, REACH, RoHS
  • Up to 10,000 hours of tubing life
Specifications
Specifications
Yes
Opaque black
Silicone
1520cm
Norprene (A 60 G)
3/8 in.
40psi
Acids
660mm
4.8mm
1.7 to 1000mL/min.
Pump Tubing
For applications requiring excellent chemical, heat, ozone and ultraviolet (UV) light resistance
8.8m
132°C
–51°C
15
Documents