Cole-Parmer™ Cole-Parmer™ Masterflex™ Standard Pump Head for Precision L/S™ tubing
Ideal for accurate and repeatable fluid transfer applications
Manufacturer:  Cole-Parmer™ 0701420
 View more versions of this product
Catalog No. MFX0701420
Description
Description
Ideal for accurate and repeatable fluid transfer applications
Specifications
Specifications
0.21 to 130mL/min.
Polycarbonate
Accurate and repeatable fluid transfer applications
3.0
40°C
0°F
Cold rolled Steel
2.5 x 2.75 x 3 in.
Pump Head
Precision L/S™ 14 tubing
Tubing loading key, single channel mounting hardware and a 15 in. L of Silicone tubing
104°F
-20°C
600rpm
Documents