Cole-Parmer™ Masterflex™ Tygon™ Fuel & Lubricant L/S™ Precision Pump Tubing
Use this Tygon™ tubing for applications involving fuels and industrial lubricants. Cole Parmer™ Masterflex™ Tygon™ Fuel & Lubricant L/S™ High Performance Precision Pump Tubing is ideal for transferring hydrocarbons, gasoline, kerosene, heating oils, cutting compounds and glycol based coolants.
Manufacturer:  Cole Parmer Instrument Co 0640116
 View more versions of this product
Catalog No. MFX0640116
Description
Description
Use this Tygon™ tubing for applications involving fuels and industrial lubricants. Cole Parmer™ Masterflex™ Tygon™ Fuel & Lubricant L/S™ High Performance Precision Pump Tubing is ideal for transferring hydrocarbons, gasoline, kerosene, heating oils, cutting compounds and glycol based coolants.
  • Tubing resists most hydrocarbons, embrittlement and swelling
  • Specially formulated to transport hydrocarbons, petroleum products and distillates
  • Thermoplastic
  • PVC-based material with plasticizer
  • Firm (stiff) material
  • Ozone- and UV-resistant
  • Low extractability
  • Translucent yellow
  • Low gas permeability
  • High dielectric constant
  • Don’t use with strong acids and alkalies and sterilization is not recommended
Specifications
Specifications
FDA 21 CFR 175.300
3.1mm
0.8 to 480mL/min.
Pump Tubing
Transferring Hydrocarbons, Gasoline, Kerosene, heating Oils, cutting compounds and Glycol based coolants
8.8m
74°C
–37°C
16
3/25 in.
Yes
Translucent Yellow
Thermoplastic PVC
1520cm
CO2: 360, H2: 97, O2: 80, N2: 40
1/8 in.
40psi
50 ft.
660mm
Documents