Oakton™ Gilmont™ Shielded Laboratory Calibrated/Correlated Flowmeters
Manufacturer:  Oakton™ GF1160
 View more versions of this product
Catalog No. 15-078-109
Includes: Calibration table for air and water and comes with glass and stainless-steel floats.
Description
Description
For micro flowmeter, accuracy is ±5% of reading or ±2 scale division for air and ±10% of reading or ±3 scale division for water.
Specifications
Specifications
125psig
±2% of the reading or ±1 scale division
1
10.25 in.
65°C
Glass and 316 Stainless Steel
Glass: 0.01 to 4; 316 Stainless: 0.02 to 8.6mL/min.
26cm
Documents