Oakton™ Gilmont™ Shielded Laboratory Calibrated/Correlated Flowmeters
Manufacturer:  Oakton™ GF1060
 View more versions of this product
Catalog No. 15-078-106
Includes: Calibration table for air and water and comes with glass and stainless-steel floats.
Description
Description
For micro flowmeter, accuracy is ±5% of reading or ±2 scale division for air and ±10% of reading or ±3 scale division for water.
Specifications
Specifications
125psig
±2% of the reading or ±1 scale division
0
10.25 in.
65°C
Glass and 316 Stainless Steel
Glass: 0.002 to 1.1; 316 Stainless: 0.004 to 2.3mL/min.
26cm
Documents