Please login to your online account to display your discounted pricing
Fisher Scientific™ FH10, FH15 and FH30 Peristaltic Tubing Pumps
Peristaltic pumps - enhanced and versatile performance in an ultra-compact, low-maintenance design
$1,536.54 - $1,711.33
Specifications
Dimensions (L x W x H) 6.5 x 5.3 x 4.5 in. (16.5 x 13.6 x 11.4cm)
Max. Pressure 2 bar
Enclosure Rating (Nema / IP) IP22
Display Green LED, On/Off
Hertz 50/60Hz
View More Specs
Products
Catalog Number Mfr. No. Housing Material Speed Flow Rate Price Quantity    
13-310-654
fisher scientific™
7777710
Painted steel 20 to 100rpm 0.8 to 105mL/min. Each for $1,536.54
13-310-653
fisher scientific™
7777704
Powder-Coated steel 300rpm 0.48 to 2.9mL/min. Each for $1,629.94
13-310-655
fisher scientific™
7777712
Painted steel 20 to 100rpm 0.8 to 14mL/min. per channel Each for $1,711.33
13-310-651
fisher scientific™
7777700
Powder-Coated steel 10rpm 0.015 to 0.08mL/min. Each for $1,629.94
Description & Specifications
Specifications
Dimensions (L x W x H) 6.5 x 5.3 x 4.5 in. (16.5 x 13.6 x 11.4cm)
Max. Pressure 2 bar
Enclosure Rating (Nema / IP) IP22
Display Green LED, On/Off
Hertz 50/60Hz
External Control-Input Start / Stop
No. of Heads Accepted 1
Certifications/Compliance UL, cUL (Power Supply) CE, RoHS, ISO9001:2008
These peristaltic pumps - provided as complete pumping systems - are ideal where space is limited. Quality design in a small, compact package, these peristaltic pumps are provided complete with pump, motor, and control in a stackable steel housing. Low-maintenance design provides thousands of hours of reliable service while meeting a wide range of fluid handling needs.
Wide Range of Performance
  • Robust, fixed occlusion design allows reliable, accurate pumping and dispensing
  • Supports wide variety of tubing materials and varying differential pressure applications
Easy-to-Use Controls
  • Mounted on front panel
  • Separate single-turn speed control
  • Flow direction switch with center OFF position
  • PRIME button runs pump at maximum speed to rapidly prime or flush tubing
  • Reversible pump direction to purge tubing
  • Remote start/stop terminal on back of unit
Easy to Maintain
  • Simple, fast tubing changes
  • Fixed occlusion eliminates adjustment after tubing changes and assures operation against pressure up to 2 bar (30psig)
High Purity Assured
  • Most tubing materials exceed USP Class VI and EP (European Pharmacopeia) standards
  • For the highest purity requirements use BioPharm silicone tubing
Manufacturer-recommended peristaltic pump tubing for FH10/FH15/FH30 pump systems