Please login to your online account to display your discounted pricing

Oakton™ Conductivity Solution


Use this conductivity solution for accurate test results in the field or lab. Oakton™ Conductivity Solutions have KCl, NaCl and 442 conversions printed on the bottle for ease of use.

Conductivity ppm NaCl: 737.1, ppm 442: 1050, ppm KCl: 757.1

Manufacturer: oakton instruments  ZZ0065315

 View more versions of this product

Catalog No. 13-300-110

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 500mL
Conductivity ppm NaCl: 737.1, ppm 442: 1050, ppm KCl: 757.1
Product Type Conductivity Standards
Chemical Name or Material Cole-Parmer Conductivity Standards

Specifications

Quantity 500mL

  • Features temperature corrections printed on the bottle
  • Certified values listed in the tables and are referenced to 25°C
  • Two-year shelf-life on all solutions