Fisherbrand™ Gel Packs
Fisherbrand Soft Gel Packs are an ideal alternative to wet or dry ice, providing reliable refrigeration for the safe transport of pharmaceuticals, and laboratory products
$49.73 - $107.40
Specifications
Color White
Material Outer Layer: Metalized PET laminated to PE, Middle Layer: Open Cell PU Foam, Inner Layer: White LDPE (FDA approved for incidental food contact).
Type Gel Pack
View More Specs
Products 4
Catalog Number Mfr. No. Size Quantity Price Quantity  
Catalog Number Mfr. No. Size Quantity Price Quantity  
07-100-112 View Documents Fisherbrand™
RS24OZGL
24 oz. 24 Case of 24 Each for $53.03
07-100-113 View Documents Fisherbrand™
RS32OZGL
32 oz. 18 Case of 18 Each for $49.73
07-100-110 View Documents Fisherbrand™
RS08OZGL
8 oz. 72 Case of 72 Each for $107.40
07-100-111 View Documents Fisherbrand™
RS16OZGL
16 oz. 36 Case of 36 Each for $65.63
Description
Description
Features:
  • Packs use a +30°F formulation
  • Non-toxic & non-caustic
  • Reusable
  • Generally used for one-way shipment
  • Low cost
  • Made in the USA
  • Solid when frozen, not flexible
  • FDA compliant
  • Gel is non-toxic, food safe
  • Place in freezer up to 72 hours before they are frozen and ready to use
  • Recycling: outer bag can be recycled, gel may be disposed as normal waste.
  • Ideal for shipments and packaging where variable height tolerances are acceptable as this formulation does tend to have irregular expansion during the freezing process.
Order Info
Case quantity only 72 per case
Warranty
Warranty against defects in material or workmanship.
Specifications
Specifications
White
Outer Layer: Metalized PET laminated to PE, Middle Layer: Open Cell PU Foam, Inner Layer: White LDPE (FDA approved for incidental food contact).
Gel Pack