Please login to your online account to display your discounted pricing

Coenzyme Q10, Tocris

Antioxidant

$95.70 - $401.94

Chemical Identifiers

Chemical Name or Material Coenzyme Q10
Assay Percent Range >99%
Molecular Formula C59H90O4
Formula Weight 863.34
Synonym 2-[(2E,6E,10E,14E,18E,22E,26E,30E,34E)-3,7,11,15,19,23,27,31,35,39-Decamethyl-2,6,10,14,18,22,26,30,34,38-tetracontadecaen-1-yl]-5,6-dimethoxy-3-methyl-2,5-cyclohexadiene-1,4-dione
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

300350

 
r & d systems
RDS300350
50mg Each for $401.94

300310

 
r & d systems
RDS300310
10mg Each for $95.70
Description & Specifications

Specifications

Formula Weight 863.34
Quantity 50mg
Molecular Formula C59H90O4
Chemical Name or Material Coenzyme Q10
CAS 303-98-0
Synonym 2-[(2E,6E,10E,14E,18E,22E,26E,30E,34E)-3,7,11,15,19,23,27,31,35,39-Decamethyl-2,6,10,14,18,22,26,30,34,38-tetracontadecaen-1-yl]-5,6-dimethoxy-3-methyl-2,5-cyclohexadiene-1,4-dione
Assay Percent Range >99%
Recommended Storage Store at -20°C
Safety and Handling

Store at -20°C