Please login to your online account to display your discounted pricing

Coenzyme A Trilithium Salt Dihydrate, 96%, Fisher BioReagents


Quantity 250mg
Packaging Amber Glass

Manufacturer: fisher scientific company  BP2510250

 View more versions of this product

Catalog No. BP2510250

  • / Each
  • / Case of 9 Each
 

Description & Specifications

Chemical Identifiers

Physical Form Powder/Solid
Quantity 250mg
Color White
Identification Pass Test
Packaging Amber Glass
Chemical Name or Material Coenzyme A
Assay Percent Range ≥96 %
CAS 18439-24-2
Purity 96% min.
OD250_OD260 0.80 to 0.89

Specifications

Chemical Name or Material Coenzyme A
CAS 18439-24-2
Assay Percent Range ≥96 %