Please login to your online account to display your discounted pricing

Coenzyme A Trilithium Salt Dihydrate, 96%, Fisher BioReagents


Quantity 1g
Packaging Amber Glass

Manufacturer: fisher scientific company  BP25101

 View more versions of this product

Catalog No. BP25101

  • / Each
  • / Case of 9 Each
 

Description & Specifications

Chemical Identifiers

Physical Form Powder/Solid
Quantity 1g
Color White
OD250_OD260 0.80 to 0.89
Chemical Name or Material Coenzyme A
Identification Pass Test
CAS 18439-24-2
Assay Percent Range ≥96 %
Purity 96% min.
Packaging Amber Glass

Specifications

Chemical Name or Material Coenzyme A
CAS 18439-24-2
Assay Percent Range ≥96 %