missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Coenzyme A Trilithium Salt Dihydrate, 96%, Fisher BioReagents
$1,608.63 - $3,000.43
Chemical Identifiers
CAS | 18439-24-2 |
---|---|
Molecular Formula | C21H33Li3N7O16P3S |
Molecular Weight (g/mol) | 785.329 |
InChI Key | QSCBPHBAFBVXRK-UHFFFAOYSA-K |
Synonym | coenzyme a, trilithium salt |
PubChem CID | 53442190 |
IUPAC Name | trilithium;[5-(6-aminopurin-9-yl)-4-hydroxy-2-[[[[3-hydroxy-2,2-dimethyl-4-oxo-4-[[3-oxo-3-(2-sulfanylethylamino)propyl]amino]butoxy]-oxidophosphoryl]oxy-oxidophosphoryl]oxymethyl]oxolan-3-yl] hydrogen phosphate |
SMILES | [Li+].[Li+].[Li+].CC(C)(COP(=O)([O-])OP(=O)([O-])OCC1C(C(C(O1)N2C=NC3=C2N=CN=C3N)O)OP(=O)(O)[O-])C(C(=O)NCCC(=O)NCCS)O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
BP2510250
|
Fisher BioReagents
BP2510250 |
250mg | Amber Glass |
Each for $1,608.63
Case of 9 Each for $14,477.67 |
|
BP2510500
|
Fisher BioReagents
BP2510500 |
500mg | Amber Glass |
Each for $3,000.43
Case of 9 Each for $27,003.87 |
|
Chemical Identifiers
18439-24-2 | |
785.329 | |
coenzyme a, trilithium salt | |
trilithium;[5-(6-aminopurin-9-yl)-4-hydroxy-2-[[[[3-hydroxy-2,2-dimethyl-4-oxo-4-[[3-oxo-3-(2-sulfanylethylamino)propyl]amino]butoxy]-oxidophosphoryl]oxy-oxidophosphoryl]oxymethyl]oxolan-3-yl] hydrogen phosphate |
C21H33Li3N7O16P3S | |
QSCBPHBAFBVXRK-UHFFFAOYSA-K | |
53442190 | |
[Li+].[Li+].[Li+].CC(C)(COP(=O)([O-])OP(=O)([O-])OCC1C(C(C(O1)N2C=NC3=C2N=CN=C3N)O)OP(=O)(O)[O-])C(C(=O)NCCC(=O)NCCS)O |
Specifications
18439-24-2 | |
Pass Test | |
C21H33Li3N7O16P3S | |
coenzyme a, trilithium salt | |
QSCBPHBAFBVXRK-UHFFFAOYSA-K | |
trilithium;[5-(6-aminopurin-9-yl)-4-hydroxy-2-[[[[3-hydroxy-2,2-dimethyl-4-oxo-4-[[3-oxo-3-(2-sulfanylethylamino)propyl]amino]butoxy]-oxidophosphoryl]oxy-oxidophosphoryl]oxymethyl]oxolan-3-yl] hydrogen phosphate | |
53442190 | |
96% min. |
White | |
≥96 % | |
Amber Glass | |
250mg | |
[Li+].[Li+].[Li+].CC(C)(COP(=O)([O-])OP(=O)([O-])OCC1C(C(C(O1)N2C=NC3=C2N=CN=C3N)O)OP(=O)(O)[O-])C(C(=O)NCCC(=O)NCCS)O | |
785.329 | |
Powder/Solid | |
Coenzyme A |