Please login to your online account to display your discounted pricing

COD Standard, 300ppm (1mL = 0.3mg COD) (Potassium Acid Phthalate in Water), Certified, 0.300mg/mL ±0.003, LabChem™

$52.09 - $52.09

Chemical Identifiers

Chemical Name or Material COD Standard
CAS 7732-18-5
CAS Max % 99.67
CAS 877-24-7
CAS Max % 0.03
CAS 7664-38-2
CAS Max % 0.3
Name Note 300ppm (1mL = 0.3mg COD) (Potassium Acid Phthalate in Water)
Purity Grade Certified Grade
Linear Formula KHC8H4O4
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC132401

 
lab chem
LC132401
Poly Bottle 500mL Each for $52.09
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 204.22
Chemical Name or Material COD Standard
Traceability to NIST Traceable to NIST
Solubility Soluble in water
Name Note 300ppm (1mL = 0.3mg COD) (Potassium Acid Phthalate in Water)
Packaging Poly Bottle
CAS Max % 0.03
Assay Percent Notes as COD
CAS Max % 99.67
CAS 877-24-7
Linear Formula KHC8H4O4
CAS 7664-38-2
CAS 7732-18-5
Concentration 0.300mg/mL ±0.003
Identification Passes Test
CAS Max % 0.3
Recommended Storage Room Temperature
Physical Form Liquid
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Quantity 500mL
Color Clear, colorless
Health Hazard 2 GHS H Statement
Solution is not hazardous.
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.

Room Temperature