Please login to your online account to display your discounted pricing

COD Standard, 1000ppm (1mL = 1mg COD) (Potassium Acid Phthalate in Water), Certified, 0.300mg/mL ±0.003, LabChem™

$42.49 - $42.49

Chemical Identifiers

Chemical Name or Material COD Standard
CAS 7732-18-5
CAS Max % 99.61
CAS 877-24-7
CAS Max % 0.09
CAS 7664-38-2
CAS Max % 0.3
Name Note 1000ppm (1mL = 1mg COD) (Potassium Acid Phthalate in Water)
Purity Grade Certified Grade
Linear Formula KHC8H4O4
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC132451

 
lab chem
LC132451
Poly Bottle 500mL Each for $42.49
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 204.22
Recommended Storage Room Temperature
Health Hazard 2 GHS H Statement
Solution is not hazardous.
Solubility Soluble in water
Quantity 500mL
Packaging Poly Bottle
CAS Max % 99.61
CAS 877-24-7
CAS 7664-38-2
CAS 7732-18-5
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Chemical Name or Material COD Standard
Assay Percent Notes as COD
Color Clear, colorless
Name Note 1000ppm (1mL = 1mg COD) (Potassium Acid Phthalate in Water)
Linear Formula KHC8H4O4
Physical Form Liquid
CAS Max % 0.09
Identification Passes Test
Concentration 0.300mg/mL ±0.003
Traceability to NIST Traceable to NIST
CAS Max % 0.3
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.

Room Temperature